Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
Specifications
| Description | CTC RTC and Thermo RSH Syringe |
| Needle Gauge | 26 G |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Product Type | Fixed Needle |
| Volume (Metric) | 10 μL |
| Needle Point Style | Bevel |
| For Use With (Equipment) | Thermo Gas Chromatography |