Learn More
Novus Biologicals™ Autophagy Antibody Pack
Product Code. 18296493
Change view
Click to view available options
Quantity:
5 Vials
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18296493 5 Vials Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18296493 Supplier Novus Biologicals™ Supplier No. NB91094877
  • ONLINE EXCLUSIVE PRICE
    7125.00 NOK
    6736.80 NOK / Pack of 6
    Save 388.20 NOK
    5% Off
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Includes
  • 1.NB100-2220: LC3B/MAP1LC3B Antibody
  • 2.NB110-53818: ATG5 Antibody
  • 3.NB110-56893: ATG9A Antibody
  • 4.NB110-82384: ATG16L1 Antibody
  • 5.NB110-87318: Beclin 1/ATG6 Antibody
For Research applications
Autophagy specifically detects Autophagy in Alligator, Bovine, Human, Mouse, Pig, Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Autophagy
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot
Concentration 25ul vials of each of following: NB100-2220, NB110-87318, NB110-53818, NB110-56893 and NB110-82384.
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation See individual datasheets
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Gene Symbols BECN1
Immunogen See individual datasheets
Quantity 5 Vials
Research Discipline Autophagy
Gene ID (Entrez) 8678
Test Specificity Autophagy Antibodies (Beclin, LC3, ATG5, ATG9, ATG16L1)
Includes 1.NB100-2220: LC3B/MAP1LC3B Antibody
2.NB110-53818: ATG5 Antibody
3.NB110-56893: ATG9A Antibody
4.NB110-82384: ATG16L1 Antibody
5.NB110-87318: Beclin 1/ATG6 Antibody
Target Species Alligator, Bovine, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.