Learn More
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Product Code. 15237626 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
50 μL
300 μL
1000 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 3840
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered Sterility Fit for Purpose Unit Size
15237626 300 μL Filtered Sterile Sterile and PCR Ready Pack of 3840
13326048 50 μL Filtered Sterile Sterile and PCR Ready Pack of 3840
15227626 50 μL Non-filtered Non-sterile Certified Quality Pack of 3840
15257626 300 μL Non-filtered Sterile Sterile Pack of 3840
15267626 300 μL Non-filtered Non-sterile Certified Quality Pack of 3840
13306068 1000 μL Filtered Sterile Sterile and PCR Ready Pack of 3840
13346068 1000 μL Non-filtered Sterile Sterile Pack of 3840
13356068 1000 μL Non-filtered Non-sterile Certified Quality Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15237626 Supplier Thermo Scientific™ Supplier No. 966021
  • ONLINE EXCLUSIVE PRICE
    9705.00 NOK
    9604.70 NOK / Pack of 3840
    Save 100.30 NOK
    1% Off
View all promotions
Estimated Shipment: 26-10-2026
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
  • Available in nonsterile, sterile and sterile filtered
  • Validated on actual workstation for which tip is created
  • Low percentage coefficient of variation
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 300 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Eppendorf epMotion
Product Line Eppendorf epMotion
Fit for Purpose Sterile and PCR Ready