Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
GREENER_CHOICE
Product Code. 11897313 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Fit for Purpose Unit Size
11897313 50 μL Sterile Filtered Sterile and PCR Ready Pack of 4800
10078172 50 μL Sterile Non-filtered Sterile Pack of 4800
10325104 50 μL Non-sterile Non-filtered Certified Quality Pack of 4800
11807323 80 μL Sterile Filtered Sterile and PCR Ready Pack of 4800
10101804 100 μL Sterile Non-filtered Sterile Pack of 4800
10654844 100 μL Non-sterile Non-filtered Certified Quality Pack of 4800
11815953 150 μL Sterile Filtered Sterile and PCR Ready Pack of 4800
10753675 200 μL Sterile Non-filtered Sterile Pack of 4800
10373653 200 μL Non-sterile Non-filtered Certified Quality Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11897313 Supplier Thermo Scientific™ Supplier No. 906021
  • ONLINE EXCLUSIVE PRICE
    8610.00 NOK
    8516.71 NOK / Pack of 4800
    Save 93.29 NOK
    1% Off
View all promotions
Estimated Shipment: 28-12-2027
Add to basket
Add to basket
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius
Product Line Tecan
Fit for Purpose Sterile and PCR Ready