Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
Description
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
- Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
- Available in filtered, low retention or extended length for increased volume to reach into deep well plates
- Lot-tested during production on actual workstation for which they are created
- Fully conductive providing increased pipetting performance
- Low percentage coefficient of variation (%CV)
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Volume (Metric) | 1000 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Black |
Autoclavable | Autoclavable |
Certifications/Compliance | ISO 9001 and ISO 13485 |
Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
Format | Rack |
Tip Style | Conductive |
Quantity | 4800 Tips |