Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
GREENER_CHOICE
Product Code. 10071541
Change view
Click to view available options
Volume (Metric):
250 μL
300 μL
1000 μL
1100 μL
Filtered:
Filtered
Non-filtered
Packaging:
96 Tips/Tray, 10 Trays/Case
96 Tips/Tray, 50 Trays/Case
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
Pack of 960
Product Code. Volume (Metric) Filtered Packaging Fit for Purpose unitSize
10071541 1100 μL Non-filtered 96 Tips/Tray, 50 Trays/Case Sterile Pack of 4800
10798302 250 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile and PCR Ready Pack of 960
10757922 250 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile and PCR Ready Pack of 960
10063411 300 μL Non-filtered 96 Tips/Tray, 50 Trays/Case Sterile Pack of 4800
10274661 300 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Sterile Pack of 960
10081001 300 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Certified Quality Pack of 960
10264711 1000 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile and PCR Ready Pack of 960
10386691 1100 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Sterile Pack of 960
10284661 1100 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Certified Quality Pack of 960
9 options
Product Code. 10071541
Brand: Thermo Scientific™ 953253
Only null left
Add to basket
Add to basket
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
  • Available in nonsterile, sterile and with the ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Conductive
Packaging 96 Tips/Tray, 50 Trays/Case
For Use With (Equipment) Universal Fit, Qiagen
Product Line Qiagen
Fit for Purpose Sterile