Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE
Product Code. 16436949 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered Sterility Fit for Purpose Unit Size
16436949 200 μL Non-filtered Sterile Sterile Pack of 4800
16406949 50 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
16416949 50 μL Non-filtered Sterile Sterile Pack of 4800
16426949 50 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11750095 80 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11986977 100 μL Non-filtered Sterile Sterile Pack of 4800
11860421 100 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11760095 150 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11806081 200 μL Non-filtered Non-sterile Certified Quality Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16436949 Supplier Thermo Scientific™ Supplier No. 923261
  • ONLINE EXCLUSIVE PRICE
    4650.00 NOK
    3444.32 NOK / Pack of 4800
    Save 1205.68 NOK
    25% Off
View all promotions
Estimated Shipment: 27-04-2027
Add to basket
Add to basket
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Sterile