Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE
Product Code. 16416949 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
Product Code. Volume (Metric) Filtered Sterility Fit for Purpose unitSize
16416949 50 μL Non-filtered Sterile Sterile Pack of 4800
16406949 50 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
16426949 50 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11750095 80 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11986977 100 μL Non-filtered Sterile Sterile Pack of 4800
11860421 100 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11760095 150 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
16436949 200 μL Non-filtered Sterile Sterile Pack of 4800
11806081 200 μL Non-filtered Non-sterile Certified Quality Pack of 4800
9 options
Product Code. 16416949
Brand: Thermo Scientific™ 920261
Only null left
Add to basket
Add to basket
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
  • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Sterile