Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Product Code. 15243059
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 4800
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Unit Size
15243059 150 μL Sterile Filtered Pack of 4800
16456949 50 μL Sterile Filtered Pack of 4800
12358592 50 μL Sterile Non-filtered Pack of 4800
16466949 50 μL Non-sterile Non-filtered Pack of 4800
16476949 80 μL Sterile Filtered Pack of 4800
16486949 100 μL Sterile Non-filtered Pack of 4800
16496949 100 μL Non-sterile Non-filtered Pack of 4800
12716027 200 μL Sterile Non-filtered Pack of 4800
16406959 200 μL Non-sterile Non-filtered Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15243059 Supplier Thermo Scientific™ Supplier No. 986021
  • ONLINE EXCLUSIVE PRICE
    8680.00 NOK
    8586.90 NOK / Pack of 4800
    Save 93.10 NOK
    1% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
  • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 150 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Low Retention
For Use With (Equipment) BioTek Pipetting System
Product Line BioTek