Learn More
- Home
- Products
- Pipette Products
- Pipette Tips
- Robotic Pipette Tips
- Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Product Code. 11741353
Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
8 μL
10 μL
20 μL
40 μL
50 μL
130 μL
175 μL
250 μL
Filtered:
Filtered
Non-filtered
Tip Style:
Extended Length
Low Retention
Standard
Wide Bore
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
Pack of 960
25 product options available for selection
| Product Code. | Volume (Metric) | Filtered | Tip Style | Fit for Purpose | unitSize |
|---|---|---|---|---|---|
| 11741353 | 50 μL | Filtered | Extended Length | Sterile | Pack of 4800 |
| 15292929 | 8 μL | Filtered | Standard | Sterile and PCR Ready | Pack of 4800 |
| 15202949 | 10 μL | Non-filtered | Standard | Sterile | Pack of 4800 |
| 15212949 | 10 μL | Non-filtered | Standard | Certified Quality | Pack of 4800 |
| 16456939 | 40 μL | Filtered | Standard | Sterile and PCR Ready | Pack of 4800 |
| 16466939 | 50 μL | Non-filtered | Standard | Sterile | Pack of 4800 |
| 15105393 | 50 μL | Non-filtered | Standard | Certified Quality | Pack of 4800 |
| 12578468 | 50 μL | Non-filtered | Low Retention | Certified Quality | Pack of 4800 |
| 11861723 | 20 μL | Filtered | Standard | Sterile and PCR Ready | Pack of 4800 |
| 11761273 | 20 μL | Filtered | Low Retention | Sterile and PCR Ready | Pack of 4800 |
| 11811733 | 20 μL | Non-filtered | Standard | Sterile | Pack of 4800 |
| 11741273 | 20 μL | Non-filtered | Low Retention | Sterile | Pack of 4800 |
| 11851723 | 20 μL | Non-filtered | Standard | Certified Quality | Pack of 4800 |
| 11751273 | 20 μL | Non-filtered | Low Retention | Certified Quality | Pack of 4800 |
| 11801733 | 130 μL | Filtered | Standard | Sterile and PCR Ready | Pack of 4800 |
| 11791273 | 130 μL | Filtered | Low Retention | Sterile and PCR Ready | Pack of 4800 |
| 16476939 | 130 μL | Filtered | Wide Bore | Sterile and PCR Ready | Pack of 4800 |
| 11881723 | 250 μL | Non-filtered | Standard | Sterile | Pack of 4800 |
| 11771273 | 250 μL | Non-filtered | Low Retention | Sterile | Pack of 4800 |
| 16486939 | 250 μL | Non-filtered | Wide Bore | Sterile | Pack of 4800 |
| 11891723 | 250 μL | Non-filtered | Standard | Certified Quality | Pack of 4800 |
| 11781273 | 250 μL | Non-filtered | Low Retention | Certified Quality | Pack of 4800 |
| 16496939 | 250 μL | Non-filtered | Wide Bore | Certified Quality | Pack of 4800 |
| 11515514 | 175 μL | Filtered | Standard | Sterile | Pack of 960 |
| 15272289 | 50 μL | Non-filtered | Standard | Certified Quality | Pack of 4800 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
Product Code.
11741353
Supplier
Thermo Scientific™
Supplier No.
2760
-
7865.00 NOK
7779.69 NOK / Pack of 4800
Save 85.31 NOK
1% Off
Estimated Shipment: 26-02-2027
Log in to see stock.
Log in to see stock.
Add to basket
7471.75 NOK valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28170" to get your promotional price.
BEST PRICE on Promo! Use promo code "28170" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
- Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384-, and 1536-deep well plates
- Low percentage coefficient of variation (%CV)
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | ISO 9001 and ISO 13485 |
| Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
| Format | Rack |
| Tip Style | Extended Length |
| Packaging | 96 tips/rack, 10 racks/pack, 5 packs/case |
| Quantity | 4800 tips |
| For Use With (Equipment) | Universal Fit |
| Product Line | Beckman |
| Fit for Purpose | Sterile |
| Show More Show Less |