Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Product Code. 11791263
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 19200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Filtered Fit for Purpose Unit Size
11791263 Sterile Non-filtered Sterile Pack of 19200
11711273 Sterile Filtered Sterile and PCR Ready Pack of 19200
11701273 Non-sterile Non-filtered Certified Quality Pack of 19200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11791263 Supplier Thermo Scientific™ Supplier No. 935261
  • ONLINE EXCLUSIVE PRICE
    75065.00 NOK
    71362.56 NOK / Pack of 19200
    Save 3702.44 NOK
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-06-2027
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile