Learn More
SymLine™ Automatic Switching Sets for Container
Product Code. 18696478
Change view
Click to view available options
For Use With (Application):
Non-explosive Atmospheres
Potentially Explosive Atmospheres
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
18696478 Non-explosive Atmospheres 1 set
18616488 Potentially Explosive Atmospheres 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18696478 Supplier SymLine™ Supplier No. 106751
  • ONLINE EXCLUSIVE PRICE
    21875.00 NOK
    19079.72 NOK / 1 sets
    Save 2795.28 NOK
    12% Off
 Restricted Item
Estimated Shipment: 04-12-2026
The ready-to-use automatic switching sets prevents the overflowing of disposal containers and the associated leakage of hazardous, toxic or explosive liquid waste.
  • Coordinated system for monitoring the safe filling of disposal containers
  • For connection to two disposal containers (not included in the set)
  • Set consisting of Built-in Signalbox2, SwitchBox, ball valve and two signal cables
  • Optionally available as an ATEX version with an additional switch amplifier for 2 channels
Specifications
Certifications/Compliance CE
Description Automatic switching set for container
Includes 1 Built-in Signalbox2, EU/UK.US, 2-channel, 2 switch inputs, 2 switch outputs, 1x SwitchBox: control unit, 1x 3-way ball valve, 3x GL25 (m), electric, 1x 3-way ball valve mount, 2x signal cable
For Use With (Application) Non-explosive Atmospheres
For Use With (Equipment) Disposal Container
Product Type Automatic Switching Set