Learn More
BRAND™ Dispenser, tilt measure without bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Description
- Dispenser head
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- Particularly well-suited for dispensing suspensions
Specifications
Specifications
Material | Borosilicate Glass 3.3 |
Description | Without bottle |
Capacity (Metric) | 5 mL |
Product Content Correction
Product Title
BRAND™ Dispenser, tilt measure without bottle >