Learn More
SI Analytics™ Automatic 3/2 Way Valve
SI Analytics™ Automatic 3/2 Way Valve is for use with wide range of products including interchangable modules.
Brand:  SI Analytics™ 285223007
Additional Details : Weight : 0.50000kg
Product Code. 10084311
  • 1735.00 NOK / Each
Estimated Shipment: 11-06-2024
Description
  • For interchangeable modules TA 05 plus through TA 50 plus and WA 05 through WA 50
  • Suitable for TitroLine™ Easy and 5000, TitroLine™ KF, TITRONIC™ Universal, Basic, and 300
Specifications
Specifications
Automatic 3-way Valve
TitroLine Alpha Plus, TitroLine KF, and TITRONIC 110 Plus