Learn More
Applied Biosystems™ AutoLys M Tube Racks
Description
• These AutoLys M Tube Racks hold 24 AutoLys M Tubes per rack
• Racks are locked and unlocked using the AutoLys M TubeLifter.
• Racks are locked and unlocked using the AutoLys M TubeLifter.
Specifications
Specifications
| Content And Storage | 5 racks (each rack holds up to 24 Tubes) |
| For Use With (Equipment) | AutoLys TubeLifter System |
| Quantity | 5 racks |
| Shipping Condition | Room Temperature |