Learn More
DuPont™ Micronova™ Autoclave Indicator Tape
Shop All Micronova Products
Click to view available options
Color:
Natural, Green, Black
Unit Size:
Each
Description
Confirms a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. DuPont™ Micronova™ Autoclave Indicator Tape adheres on contact and stays in place through live steam pressure. After use, the tape removes quickly and completely, leaving minimal residue or staining.
- Natural background colour, with printed green indicator lines, changing to black when proper levels of steam pressure and temperature have been achieved.
- Uniquely printed lines cannot be rubbed off and will not contaminate instruments.
- Saturated crepe paper backing
- Synthetic polyisoprene adhesive
- Lead-free ink indicator
- Standard sterilisation cycle (with pre-vacuum cycle): 121°C (250°F) for 20 minutes, 132° to 134°C (270 to 274°F) for 4 minutes
- 25mm x 55m length, wound onto a plastic core.
Specifications
Specifications
| Adhesive Type | Polyisoprene, Synthetic |
| Color | Natural, Green, Black |
| Format | Tape |
| Material | Crepe Paper |
| Sterility | Sterile |
| For Use With (Equipment) | Gravity discharge and vacuum autoclaves |
| Quantity | 1 Ea. |
| Dimensions (LxW) | 55 cm x 25.4 cm |
| Length (English) | 2.16 in. |
| Length (Metric) | 55 mm |
| Show More |