Learn More
- Home
- Products
- Sterilizers and Autoclaves
- Autoclave Bags, Wraps, and Sleeves
- Keystone Cleanroom Products Autoclavable Tyvek™ Breather Bags with Self-Sealing Closure
Keystone Cleanroom Products Autoclavable Tyvek™ Breather Bags with Self-Sealing Closure
Product Code. 30308739
Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Length (English):
6 in.
8 in.
9 in.
10 in.
11 in.
12 in.
13 in.
14 in.
15 in.
17 in.
18 in.
20 in.
22 in.
24 in.
26 in.
28 in.
30 in.
32 in.
34 in.
35 in.
Width (English):
2 in.
3 in.
3.5 in.
4 in.
5 in.
6 in.
7 in.
8 in.
10 in.
12 in.
14 in.
15 in.
16 in.
18 in.
24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
32 product options available for selection
| Product Code. | Length (English) | Width (English) | Unit Size |
|---|---|---|---|
| 30308739 | 35 in. | 24 in. | Pack of 250 |
| 30308707 | 6 in. | 2 in. | Pack of 1000 |
| 30308682 | 9 in. | 2 in. | Pack of 1000 |
| 30308720 | 6 in. | 3 in. | Pack of 1000 |
| 30308657 | 8 in. | 3.5 in. | Pack of 500 |
| 30308665 | 15 in. | 4 in. | Pack of 500 |
| 30308692 | 22 in. | 4 in. | Pack of 500 |
| 30308661 | 10 in. | 5 in. | Pack of 500 |
| 30308695 | 8 in. | 6 in. | Pack of 500 |
| 30308708 | 12 in. | 6 in. | Pack of 500 |
| 30308669 | 28 in. | 6 in. | Pack of 500 |
| 30308711 | 32 in. | 6 in. | Pack of 500 |
| 30308738 | 35 in. | 6 in. | Pack of 500 |
| 30308735 | 13 in. | 7 in. | Pack of 500 |
| 30308712 | 10 in. | 8 in. | Pack of 500 |
| 30308654 | 15 in. | 8 in. | Pack of 500 |
| 30308704 | 18 in. | 8 in. | Pack of 500 |
| 30308655 | 34 in. | 8 in. | Pack of 500 |
| 30308677 | 11 in. | 10 in. | Pack of 500 |
| 30308688 | 20 in. | 10 in. | Pack of 500 |
| 30308721 | 14 in. | 12 in. | Pack of 500 |
| 30308722 | 17 in. | 12 in. | Pack of 500 |
| 30308734 | 24 in. | 14 in. | Pack of 500 |
| 30308678 | 20 in. | 15 in. | Pack of 500 |
| 30308689 | 17 in. | 16 in. | Pack of 500 |
| 30308736 | 26 in. | 16 in. | Pack of 500 |
| 30308691 | 32 in. | 16 in. | Pack of 250 |
| 30308696 | 35 in. | 16 in. | Pack of 250 |
| 30308693 | 22 in. | 18 in. | Pack of 500 |
| 30308670 | 26 in. | 24 in. | Pack of 500 |
| 30308667 | 30 in. | 24 in. | Pack of 250 |
| 30308684 | 8 in. | 3 in. | Pack of 500 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
The medical grade 1073B Tyvek™ material has the highest microbial barrier properties of any steam sterilization compatible materials. Tyvek is also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
- Manufactured and packaged in an ISO certified cleanroom
- Chevron Peel Opening
- Autoclavable at up to 125°C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
- Convenient Self-Sealing Closure
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Self-seal |
| Color | Clear, White |
| Disposable | Single-use |
| For Use With (Application) | Steam, EtO, Gamma or VHP Sterilization |
| Material | Tyvek |
| Sterility | Non-sterile |
| Latex Free | Yes |
| Dimensions (L x W) | 24 x 35 in. |
| Length (English) | 35 in. |
| Width (English) | 24 in. |
| Product Type | Autoclave Bag |
| Length (Metric) | 88.9 cm |
| Width (Metric) | 61 cm |
| Show More Show Less |