Learn More
Cole-Parmer™ PVDF Compression to Male Threaded Adapter Fittings
Product Code. 11754749
Click to view available options
For Use With:
1/4 tubing O.D. x 1/4 in. NPT(M) Thread
1/4 tubing O.D. x 1/8 in. NPT(M) Thread
3/8 tubing O.D. x 1/4 in. NPT(M) Thread
3/8 tubing O.D. x 3/8 in. NPT(M) Thread
Port Size:
11/32 in. orifice, 1-13/32 x 13/16 in.
5/16 in. orifice, 1-11/32 x 13/16 in.
7/32 in. orifice, 1-3/16 x 5/8 in.
7/32 in. orifice; 1 x 5/8 in.
Thread Size:
1/4 in. NPT(M)
1/8 in. NPT(M)
3/8 in. NPT(M)
Unit Size:
Each
Product Code. 11754749
Brand: Cole-Parmer™ 0638584
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These compression fittings are an excellent choice to connect rigid tubing systems, with higher pressure resistance than that provided by barbed fittings.
  • For rigid tubing systems, including tubing made from fluoropolymers, other rigid plastics, and metals
  • Assemble the fitting to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • No leaks, flow restrictions and costly downtime
  • Sterilization by autoclave or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PVDF
For Use With 3/8 tubing O.D. x 1/4 in. NPT(M) Thread
Packaging Quantity 1 Each
Port Size 5/16 in. orifice, 1-11/32 x 13/16 in.
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/4 in. NPT(M)
Product Type Compression Fitting