Learn More
Masterflex™ Celcon™ Ratchet Tubing Clamps
Description
Twelve-position ratchet control adjusts from full flow to complete closure. Autoclavable.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Quantity | 25/Pk. |
| Type | Tubing Clamp |
| For Use With (Equipment) | For up to 1/2 in. tubing |