Learn More
Cole-Parmer™ Elbow Fitting, Compression x NPT(M), PFA
Product Code. 11731989
Click to view available options
Tubing Size:
1/4 in. OD Tubing, 1/8 in. NPT
Unit Size:
Each
Product Code. 11731989
Brand: Cole-Parmer™ 3132015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cole-Parmer™ Elbow Fitting, Compression x NPT(M), PFA features threaded and compression fittings for a reliable and secure connection.
  • Easy to use
  • PFA construction
  • Reliable threaded and compression type connections
Specifications
Autoclavable Autoclavable
For Use With 1/4 OD tubing and 1/8 in. NPT thread connection
Packaging Quantity 1 Each
Tubing Size 1/4 in. OD Tubing, 1/8 in. NPT
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/8 in. NPT