Learn More
Cole-Parmer™ Parker Hannifin™ PFA Threaded Adapter Compression Fitting
Product Code. 11795728
Click to view available options
For Use With:
1/2 tubing O.D. x 1/2 in. NPT(M) Thread
1/4 tubing O.D. x 1/4 in. NPT(F) Thread
1/4 tubing O.D. x 1/8 in. NPT(M) Thread
Thread Size:
1/2 in. NPT(M)
1/4 in. NPT(F)
1/8 in. NPT(M)
Unit Size:
Each
Product Code. 11795728
Brand: Cole-Parmer™ 3132008
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and use the nut to compress down to form a secure seal
  • Superior chemical resistance with durable PFA
  • Available in male and female varieties
  • Sterilization through autoclave or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PFA
For Use With 1/4 tubing O.D. x 1/8 in. NPT(M) Thread
Packaging Quantity 1 Each
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/8 in. NPT(M)
Product Type Compression Fitting