Learn More
Abeba™ Autoclavable Clogs 9620
Product Code. 15643268
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. Size unitSize
15643268 43 1 pair
15673258 36 1 pair
15683258 37 1 pair
15693258 38 1 pair
15603268 39 1 pair
15613268 40 1 pair
15623268 41 1 pair
15633268 42 1 pair
15653268 44 1 pair
15663268 45 1 pair
15673268 46 1 pair
11 options
Product Code. 15643268
Brand: Abeba™ 9620/43
Only null left
Add to basket
Add to basket
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Green
Size 43
Style 9620