Learn More
Abeba™ Autoclavable Clogs 9600
Product Code. 15670456
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15670456
Brand: Abeba™ 9600/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • White
  • Slip resistance outsole sole
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color White
Size 36
Sole Features Slip-resistant
Style 9600