Learn More
Abeba™ Autoclavable Clogs 39620
Product Code. 15651258
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15651258 39 1 pair
15621258 36 1 pair
15631258 37 1 pair
15641258 38 1 pair
15661258 40 1 pair
15671258 41 1 pair
15681258 42 1 pair
15691258 43 1 pair
15601268 44 1 pair
15611268 45 1 pair
15621268 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15651258 Supplier Abeba™ Supplier No. 39620/39
  • ONLINE EXCLUSIVE PRICE
    647.00 NOK
    572.09 NOK / 1 pairs
    Save 74.91 NOK
    11% Off
Estimated Shipment: 26-10-2026
Antistatic and autoclavable at 135°C for 5 minutes. High slip resistance. Washing at 60°C.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Green
Size 39
Sole Features Slip-resistant
Style 39620