Learn More
Abeba™ Autoclavable Clogs 39600
Product Code. 15651238
Change view
Click to view available options
Size:
36
38
39
40
41
42
44
45
46
Unit Size:
1 pair
Product Code. Size unitSize
15651238 41 1 pair
15601238 36 1 pair
15621238 38 1 pair
15631238 39 1 pair
15641238 40 1 pair
15661238 42 1 pair
15681238 44 1 pair
15691238 45 1 pair
15601248 46 1 pair
9 options
Product Code. 15651238
Brand: Abeba™ 39600/41
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • White
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color White
Size 41
Sole Features Slip-resistant
Style 39600