missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Auramine O, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Formula Weight | 303.8 |
| Color | Yellow |
| Quantity | 50 g |
| Physical Form | Powder |
| Chemical Name or Material | Auramine O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction