Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Product Code. p-8006555
Click to view available options
Filter Type:
FFP2
FFP3
Unit Size:
Pack of 10
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12932505
Brand: 3M™ 7000088835
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
  • Provide reliable and comfortable protection from a range of airborne disposable hazards
  • Minimize contamination risks to food or pharmaceutical production processes
  • Made without staples or small detachable parts
  • Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Color Blue
Filter Type FFP2
With Exhalation Valve Yes
Filter Efficiency FFP2
For Use With (Application) Exposure to airborne duest particles and non-volatile liquid particles
Includes Cool Flow™ exhalation valve
Material Headstrap Thermoplastic Elastomer
Nose Piece Material Aluminum
Resistant To Airborne dust and non-volatile liquid particles
Sterility Non-sterile
FDA Cleared Surgical Mask No