Learn More
Invitrogen™ Attune™ Xenith™ Bottle Caps
Product Code. 31759233
Change view
Click to view available options
Product Type:
Bleach Bottle Cap
DI Water Bottle Cap
Focus Cube Cap
Shutdown Bottle Cap
Wash Bottle Cap
Waste Cube Cap
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31759233 Wash Bottle Cap Each
31759193 Focus Cube Cap Each
31759144 Waste Cube Cap Each
31759224 Bleach Bottle Cap Each
31759214 Shutdown Bottle Cap Each
31759155 DI Water Bottle Cap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31759233 Supplier Invitrogen™ Supplier No. 100120478CS
Estimated Shipment: 16-10-2026
These parts are replacement bottle caps for the containers on the fluidics cart of the Attune Xenith Flow Cytometer.
The Invitrogen Attune Xenith Flow Cytometer has an external fluid cart that holds the fluids required for everyday use of the system. These parts are replacement bottle caps for the cart’s fluidics containers, offered here in the event that damage occurs to the bottle caps provided with the instrument.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type Wash Bottle Cap