Learn More
Attune™ NxT Wash Bottle
Description
This is a 175 mL replacement bottle for wash solution for the Attune™ NxT Acoustic Focusing Cytometer.
Specifications
Specifications
| Content And Storage | Room temperature |
| Quantity | 1 Each |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Attune™ NxT Wash Bottle |