Learn More
Invitrogen™ Attune™ NxT Violet 6 Conversion Kit
Product Code. 31722922
Change view
Click to view available options
Color:
Red
Violet
Yellow
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31722922 Red 1 set
31722852 Violet 1 set
31722771 Yellow 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31722922 Supplier Invitrogen™ Supplier No. A36571
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.
Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage Optical block and filters
Quantity 1 kit
Color Red
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT Four-Channel Violet Instrument
Product Type Conversion Kit