Learn More
Invitrogen™ Attune™ NxT Violet 6 Conversion Kit
Description
Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.
Specifications
Specifications
| Product Line | Attune |
| Content And Storage | Optical block and filters |
| Quantity | 1 kit |
| Color | Violet |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Attune NxT Four-Channel Violet Instrument |
| Product Type | Conversion Kit |