Learn More
Invitrogen™ Attune™ NxT and CytPix Sample Syringe
Product Code. 15328696
Change view
Click to view available options
Quantity:
1 Syringe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15328696 1 Syringe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15328696 Supplier Invitrogen™ Supplier No. 100022591
  • ONLINE EXCLUSIVE PRICE
    1316.00 NOK
    1210.65 NOK / Pack of 1
    Save 105.35 NOK
    8% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This 1 mL sample syringe is used with the fluidics system of the Attune™ NxT Acoustic Focusing Cytometer to move sample to the flow cell.
This 1 mL sample syringe is used with the fluidics system of the Attune™ NxT Acoustic Focusing Cytometer to move sample to the flow cell. Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 Syringe
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Sampler Syringe