Learn More
Attune™ NxT Focusing Fluid Bottle
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Description
This is a 1.9 L replacement bottle for the focusing fluid tank on the Attune™ NxT Acoustic Focusing Cytometer.
Specifications
Specifications
| Content And Storage | Room temperature |
| Quantity | 1 Each |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Attune™ NxT Focusing Fluid Bottle |