Specifications
| Product Line | Attune |
| Content And Storage | Dell 64 bit computer with Windows 7 operating system, Attune™ Cytometric Software V2.1 and V2.1 documentation Kit, and a visit by a trained Service personnel assist in data management from the existing computer system |
| Quantity | 1 kit |
| Software Type | Attune™ Cytometric Software |
| Operating System | Windows 7 (Service Pack 2) |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Computer Upgrade Kit |