Learn More
Applied Biosystems™ Attune™ Flow Cytometer Syringe
Product Code. 15830413 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830413 1 mL 1mL
17246493 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15830413 Supplier Applied Biosystems™ Supplier No. 4452079
 Restricted Item
Estimated Shipment: 16-10-2026
The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage Store at room temperature
Quantity 1 mL
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Syringe