Learn More
3M™ Attest™ Vaporized Hydrogen Peroxide Tri-Metric Chemical Indicator Strips
Descripción
Upon exposure to vaporized hydrogen peroxide, the color of the coated indicator strip progressively changes from blue toward pink. The progression of the color change along the strip is visible through a window with marked 'REJECT' and 'ACCEPT' zones.
- The first FDA cleared CI for VH2O2 sterilization, categorized by the FDA as a multivariable CI for chemical vapor sterilization that independently monitors the three critical variables of time, temperature and VH2O2 concentration
- Accurately detects failed VH2O2 sterilization cycles
- Won't change color before sterilization under ambient light
- Consistent blue-toward-pink color change for all 3M™ chemical indicators for vaporized hydrogen peroxide
- Proprietary protective film prevents ink smear on surgical instruments
Especificaciones
Especificaciones
| Certifications/Compliance | FDA VH2O2 Sterilization |
| For Use With (Equipment) | Wrapped Trays, Rigid Containers, Sterilization Pouches, Packs |
| Length (English) | 2 in. |
| Length (Metric) | 5.1 cm |
| Model | 1348 |
| Product Type | Indicator Strip |
| Quantity | 25 Strips/Pouch, 4 Pouches/Cs. |
| Sterility | Sterile |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?