Learn More
WTW™ Attachment Stirrer for WTW™ Oxygen Probe
GREENER_CHOICE
Product Code. 10099260
Change view
Click to view available options
For Use With (Equipment):
Oxygen probe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10099260 Oxygen probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10099260 Supplier WTW™ Supplier No. 203824
In Stock
Specifications
Type Attachment Stirrer
For Use With (Application) Provides a constant, defined flow of the sensor, in conjunction with the Oxi-Stirrer 300/magnetic stirrer
For Use With (Equipment) Oxygen probe