Learn More
Abnova™ ATRX/CENXp FISH Probe
Product Code. 16023375
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 uL
200 uL
missing translation for 'unitSize'
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16023375 100 uL 100µL
16033375 200 uL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 16023375 missing translation for 'mfr' Abnova™ missing translation for 'supplierNo' FG0228.100uL
  • ONLINE EXCLUSIVE PRICE
    20550.00 NOK
    19713.21 NOK / 100µL
    Save 836.79 NOK
    4% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
Quantity 100 uL
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe