missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1F Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. p-7207630 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857674 20 μL 20µL
16847674 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857674

Brand: Proteintech 177251AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.

Specifications

Antigen ATP6V1F
Applications Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP6V1F
Gene Accession No. Q16864, Q9D1K2
Gene Alias ATP6S14, ATP6V1F, V ATPase 14 kDa subunit, V ATPase subunit F, V type proton ATPase subunit F, Vacuolar proton pump subunit F, VATF, Vma7
Gene Symbols Atp6v1f
Host Species Rabbit
Immunogen ATP6V1F Fusion Protein Ag12121
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66144, 9296
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.