Learn More
Abnova™ ATP6V1B1 (Human) Recombinant Protein
Human ATP6V1B1 full-length ORF ( AAH63411, 1 a.a. - 513 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000525-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQRSGQVLEVAGTKAIVQVFEGTSGIDARKTTCEFTGDILRTPVSEDMLGRVFNGSGKPIDKGPVVMAEDFLDINGQPINPHSRIYPEEMIQTGISPIDVMNSIARGQKIPIFSAAGLPHNEIAAQICRQAGLVKKSKAVLDYHDDNFAIVFAAMGVNMETARFFKSDFEQNGTMGNVCLFLNLANDPTIERIITPRLALTTAEFLAYQCEKHVLVILTDMSSYAEALREVSAAREEVPGRRGFPGYMYTDLATIYERAGRVEGRGGSITQIPILTMPNDDITHPIPDLTGFITEGQIYVDRQLHNRQIYPPINVLPSLSRLMKSAIGEGMTRKDHGDVSNQLYACYAIGKDVQAMKAVVGEEALTSEDLLYLEFLQKFEKNFINQGPYENRSVFESLDLGWKLLRIFPKEMLKRIPQAVIDEFYSREGALQDLAPDTAL
Specifications
AAH63411 | |
ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6V1B1 | |
GST |
525 | |
Wheat germ expression system | |
25 μg | |
ATP6B1, MGC32642, RTA1B, VATB, VMA2, VPP3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |