Learn More
Abnova™ ATP5O (Human) Recombinant Protein
Product Code. p-7163992
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16112091
Brand: Abnova™ H00000539P01.10ug
Human ATP5O full-length ORF ( AAH21233, 1 a.a. - 213 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAAPAVSGLSRQVRCLSTSVVRPFAKLVRPPVQVYGIEGRYATALYSAASKQNKLEQVEKELLRVAQILKEPKVAASVLNPYVKRSIKVKSLNDITAKERFSPLTTNLINLLAENGRLSNTQGVVSAFSTMMSVHRGEVPCTVTSASPLEEATLSELKTVLKSFLSQGQVLKLEAKTDPSILGGMIVRIGEKYVDMSVKTKIQKLGRAMREIV
Specifications
Accession Number AAH21233
Gene ID (Entrez) 539
Name ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ATPO, OSCP
Gene Symbol ATP5O
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer