missing translation for 'onlineSavingsMsg'
Learn More

ATP5J2 Mouse anti-Human, Mouse, Rabbit, Rat, Clone: 2B3C3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68128-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17731189

  • 3828.53 NOK / 150µL
Estimated Shipment: 29-09-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATP5J2 Monoclonal antibody specifically detects ATP5J2 in Human, Mouse, Rabbit, Rat samples. It is validated for WB, ELISA
Specifications

Specifications

ATP5J2
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Mouse
150 μL
9551
Store at -20°C
Liquid
Western Blot, ELISA
2B3C3
Unconjugated
BC014073
Protein G purification
Primary
Human, Mouse, Rabbit, Rat
Antibody
IgG1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers