missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATP5I (Human) Recombinant Protein
Human ATP5I full-length ORF ( AAH03679, 1 a.a. - 69 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000521-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVPPVQVSPLIKLGRYSALFLGVAYGATRYNYLKPRAEEERRIAAEEKKKQDELKRIARELAEDDSILK
Specifications
AAH03679 | |
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP5I | |
GST |
521 | |
Wheat germ expression system | |
25 μg | |
ATP5K, MGC12532 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |