missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATP1B3 (Human) Recombinant Protein
Human ATP1B3 full-length ORF ( AAH11835, 1 a.a. - 279 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000483-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTKNEKKSLNQSLAEWKLFIYNPTTGEFLGRTAKSWGLILLFYLVFYGFLAALFSFTMWVMLQTLNDEVPKYRDQIPSPGLMVFPKPVTALEYTFSRSDPTSYAGYIEDLKKFLKPYTLEEQKNLTVCPDGALFEQKGPVYVACQFPISLLQACSGMNDPDFGYSQGNPCILVKMNRIIGLKPEGVPRIDCVSKNEDIPNVAVYPHNGMIDLKYFPYYGKKLHVGYLQPLVAVQVSFAPNNTGKEVTVECKIDGSANLKSQDDRDKFLGRVMFKITARA
Specifications
AAH11835 | |
ATPase, Na+/K+ transporting, beta 3 polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP1B3 | |
GST |
483 | |
Wheat germ expression system | |
10 μg | |
ATPB-3, CD298, FLJ29027 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |