Learn More
ATP Solution (10 mM)
Product Code. 10023013
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10023013 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10023013 Supplier Invitrogen Supplier No. PV3227
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
ATP Solution (10 mM)
ATP is an essential component for kinase assays using LanthaScreen™ Technology. Invitrogen's ATP is 10mM in water.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ATP is shipped on dry ice and should be stored at -20°C.
Assay Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity
Shipping Condition Dry Ice
Concentration 10 mM
Label or Dye Unlabeled
Quantity 500 μL