Learn More
MedChemExpress ATP
Specifications
Specifications
| Molecular Weight (g/mol) | 507.18 g/mol |
| Product Type | Natural Products |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.0% |
| Quantity | 100 mg |
| Solubility Information | H2O : ≥ 100 mg/mL (197.17 mM) |
| For Use With (Application) | COVID-19-immunoregulation |
| CAS | 56-65-5 |
| Synonym | Adenosine 5'-triphosphate |