missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CD49b antibody from Proteintech is a armenian hamster monoclonal antibody generated with mouse colon carcinoma cell line colon26 of mouse CD49b. This antibody recognizes mouse antigen. The CD49b antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.
Specifications
Specifications
| Antigen | CD49b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HMA2 |
| Concentration | 500 μg/mL |
| Conjugate | Atlantic Blue |
| Host Species | Armenian Hamster |
| Immunogen | Mouse colon carcinoma cell line Colon26 |
| Purification Method | Affinity purification |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?