missing translation for 'onlineSavingsMsg'
Learn More

Aticaprant, MedChemExpress

Product Code. 16490869
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
50 mg
Unit Size:
10mg
1mg
20mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490869

Brand: Medchem Express HY101718

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Aticaprant (CERC-501) is a potent and centrally-penetrant kappa opioid receptor antagonist with a Ki of 0.807 nM.

Aticaprant (CERC-501) is a potent and centrally-penetrant kappa opioid receptor antagonist with a Ki of 0.807 nM.

Specifications

Chemical Name or Material Aticaprant
CAS 1174130-61-0
Color White
Purity Grade Notes Research
Molecular Formula C26H27FN2O2
Quantity 1 mg
Synonym CERC-501 LY-2456302
Solubility Information DMSO : ≥ 100 mg/mL (238.95 mM)
SMILES O=C(N)C1=CC=C(OC2=CC=C(CN3[C@H](C4=CC(C)=CC(C)=C4)CCC3)C=C2)C(F)=C1
Molecular Weight (g/mol) 418.5
Formula Weight 418.5
Percent Purity 98.45%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.