Learn More
uvex Athletic Lite ESD Safety Glove
Product Code. p-7228493 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
Product Code. Size unitSize
17707569 6 10 pairs
17717569 7 10 pairs
17727569 8 10 pairs
17737569 9 10 pairs
17747569 10 10 pairs
17757569 11 10 pairs
6 options
Product Code. 17707569
Brand: uvex 6003506
Only null left
Add to basket
Add to basket
Particularly lightweight safety glove with ESD and touchscreen function.
  • Light and breathable safety glove
  • Particularly flexible and sensitive glove for multiple areas of application
  • Touchscreen capability
  • Certified in line with Oeko-Tex Standard 100
  • Good grip in dry and slightly damp areas
  • High breathability thanks to open-pored coating that reduces perspiration inside the glove
  • Good tactile feel when carrying out precision work
  • Perfect fit thanks to the 'slim fit' design and elastane in the liner
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN 16350, EN 388 (4121X)
Coating Material NBR MicroFoam
Color Blue, Anthracite
Disposable Reusable
Exterior Finish Smooth
Hand Ambidextrous
Laboratory/Cleanroom Use Not tested
Latex Free Yes
Material Polyamide, Carbon Fiber, Elastane
Powdered Powder Free
Product Type ESD Gloves
Size 6