Learn More
ATG9A Mouse anti-Human, Unconjugated, Clone: 1D12E11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67096-1-IG-150UL
This item is not returnable.
View return policy
Description
The ATG9A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATG9A. This antibody recognizes human antigen. The ATG9A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| ATG9A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1D12E11 | |
| Unconjugated | |
| 150 μL | |
| 79065 | |
| IgG1 |